N-(5-Amino-2-fluorophenyl)tetrahydro-2-furancarboxamide

Supplier: Matrix Scientific

049012-500MG 049012-2.5G
101885-952EA 364.62 USD
101885-952 101885-954
N-(5-Amino-2-fluorophenyl)tetrahydro-2-furancarboxamide
N-(5-Amino-2-fluorophenyl)tetrahydro-2-furancarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃FN₂O₂
MDL Number: MFCD09050471
CAS Number: 926261-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR