N-(3-Amino-4-methylphenyl)tetrahydro-2-furancarboxamide

Supplier: Matrix Scientific

049011-500MG 049011-2.5G
101885-948EA 364.62 USD
101885-948 101885-950
N-(3-Amino-4-methylphenyl)tetrahydro-2-furancarboxamide
N-(3-Amino-4-methylphenyl)tetrahydro-2-furancarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD09931673
CAS Number: 1016719-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR