N-(3-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide

Supplier: Matrix Scientific

049009-500MG 049009-2.5G
101885-940EA 364.62 USD
101885-940 101885-942
N-(3-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide
N-(3-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD09050692
CAS Number: 926215-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR