N-(4-Amino-2-methoxyphenyl)nicotinamide

Supplier: Matrix Scientific

049006-500MG 049006-2.5G
101885-928EA 364.62 USD
101885-928 101885-930
N-(4-Amino-2-methoxyphenyl)nicotinamide
N-(4-Amino-2-methoxyphenyl)nicotinamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂
MDL Number: MFCD02585685
CAS Number: 552814-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR