N-(3-Amino-4-chlorophenyl)nicotinamide

Supplier: Matrix Scientific

049003-500MG 049003-2.5G
101885-920EA 364.62 USD
101885-920 101885-922
N-(3-Amino-4-chlorophenyl)nicotinamide
N-(3-Amino-4-chlorophenyl)nicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O
MDL Number: MFCD09045269
CAS Number: 926255-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR