N-(5-Amino-2-chlorophenyl)nicotinamide

Supplier: Matrix Scientific

049002-500MG 049002-2.5G
101885-916EA 364.62 USD
101885-916 101885-918
N-(5-Amino-2-chlorophenyl)nicotinamide
N-(5-Amino-2-chlorophenyl)nicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O
MDL Number: MFCD11568692
CAS Number: 937392-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR