N-(5-Amino-2-methoxyphenyl)nicotinamide

Supplier: Matrix Scientific

049001-500MG 049001-2.5G
101885-912EA 364.62 USD
101885-912 101885-914
N-(5-Amino-2-methoxyphenyl)nicotinamide
N-(5-Amino-2-methoxyphenyl)nicotinamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂
MDL Number: MFCD09045111
CAS Number: 926204-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR