N-(3-Amino-4-fluorophenyl)nicotinamide

Supplier: Matrix Scientific

049000-500MG 049000-2.5G
101885-908EA 364.62 USD
101885-908 101885-910
N-(3-Amino-4-fluorophenyl)nicotinamide
N-(3-Amino-4-fluorophenyl)nicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀FN₃O
MDL Number: MFCD09045963
CAS Number: 926241-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR