N-(5-Amino-2-fluorophenyl)nicotinamide

Supplier: Matrix Scientific

048999-500MG 048999-2.5G
101885-904EA 364.62 USD
101885-904 101885-906
N-(5-Amino-2-fluorophenyl)nicotinamide
N-(5-Amino-2-fluorophenyl)nicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀FN₃O
MDL Number: MFCD09049330
CAS Number: 926241-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR