N-(3-Amino-4-methylphenyl)nicotinamide

Supplier: Matrix Scientific

048998-500MG 048998-2.5G
101885-900EA 364.62 USD
101885-900 101885-902
N-(3-Amino-4-methylphenyl)nicotinamide
N-(3-Amino-4-methylphenyl)nicotinamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O
MDL Number: MFCD02585704
CAS Number: 653584-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR