N-(4-Amino-2-methoxyphenyl)benzamide

Supplier: Matrix Scientific

048988-500MG 048988-2.5G
101885-860EA 364.62 USD
101885-860 101885-862
N-(4-Amino-2-methoxyphenyl)benzamide
N-(4-Amino-2-methoxyphenyl)benzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD03166488
CAS Number: 104478-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR