N-(5-Amino-2-chlorophenyl)benzamide

Supplier: Matrix Scientific

048987-500MG 048987-2.5G
101885-856EA 364.62 USD
101885-856 101885-858
N-(5-Amino-2-chlorophenyl)benzamide
N-(5-Amino-2-chlorophenyl)benzamide

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂O
MDL Number: MFCD01145120
CAS Number: 293737-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR