N-(4-Amino-2-methoxyphenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048970-500MG 048970-2.5G
101885-788EA 364.62 USD
101885-788 101885-790
N-(4-Amino-2-methoxyphenyl)cyclobutanecarboxamide
N-(4-Amino-2-methoxyphenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD09043953
CAS Number: 312303-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR