N-(4-Amino-2-methylphenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048969-500MG 048969-2.5G
101885-784EA 364.62 USD
101885-784 101885-786
N-(4-Amino-2-methylphenyl)cyclobutanecarboxamide
N-(4-Amino-2-methylphenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD09045465
CAS Number: 926238-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR