N-(3-Amino-4-chlorophenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048967-500MG 048967-2.5G
101885-776EA 364.62 USD
101885-776 101885-778
N-(3-Amino-4-chlorophenyl)cyclobutanecarboxamide
N-(3-Amino-4-chlorophenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O
MDL Number: MFCD09046433
CAS Number: 916052-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR