N-(5-Amino-2-chlorophenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048966-500MG 048966-2.5G
101885-772EA 364.62 USD
101885-772 101885-774
N-(5-Amino-2-chlorophenyl)cyclobutanecarboxamide
N-(5-Amino-2-chlorophenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O
MDL Number: MFCD12135646
CAS Number: 1155532-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR