N-(3-Amino-4-fluorophenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048964-500MG 048964-2.5G
101885-764EA 364.62 USD
101885-764 101885-766
N-(3-Amino-4-fluorophenyl)cyclobutanecarboxamide
N-(3-Amino-4-fluorophenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃FN₂O
MDL Number: MFCD09049983
CAS Number: 926258-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR