N-(5-Amino-2-fluorophenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048963-500MG 048963-2.5G
101885-760EA 364.62 USD
101885-760 101885-762
N-(5-Amino-2-fluorophenyl)cyclobutanecarboxamide
N-(5-Amino-2-fluorophenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃FN₂O
MDL Number: MFCD09045568
CAS Number: 926255-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR