N-(3-Amino-4-methylphenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048962-500MG 048962-2.5G
101885-756EA 364.62 USD
101885-756 101885-758
N-(3-Amino-4-methylphenyl)cyclobutanecarboxamide
N-(3-Amino-4-methylphenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD09930575
CAS Number: 1016506-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR