N-(5-Amino-2-methylphenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048961-500MG 048961-2.5G
101885-752EA 364.62 USD
101885-752 101885-754
N-(5-Amino-2-methylphenyl)cyclobutanecarboxamide
N-(5-Amino-2-methylphenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD09044855
CAS Number: 926223-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR