N-(4-Amino-2-methoxyphenyl)cyclopropanecarboxamide

Supplier: Matrix Scientific

048957-500MG 048957-2.5G
101885-736EA 364.62 USD
101885-736 101885-738
N-(4-Amino-2-methoxyphenyl)cyclopropanecarboxamide
N-(4-Amino-2-methoxyphenyl)cyclopropanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD09050906
CAS Number: 926231-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR