N-(3-Amino-4-fluorophenyl)cyclopropanecarboxamide

Supplier: Matrix Scientific

048955-500MG 048955-2.5G
101885-728EA 364.62 USD
101885-728 101885-730
N-(3-Amino-4-fluorophenyl)cyclopropanecarboxamide
N-(3-Amino-4-fluorophenyl)cyclopropanecarboxamide

Hazard: Irritant

Formula: C₁₀H₁₁FN₂O
MDL Number: MFCD09046486
CAS Number: 926210-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR