N-(4-Amino-2-methoxyphenyl)-3-methylbutanamide

Supplier: Matrix Scientific

048952-500MG 048952-2.5G
101885-716EA 364.62 USD
101885-716 101885-718
N-(4-Amino-2-methoxyphenyl)-3-methylbutanamide
N-(4-Amino-2-methoxyphenyl)-3-methylbutanamide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD03906877
CAS Number: 760204-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR