N-(3-Amino-4-fluorophenyl)-3-methylbutanamide

Supplier: Matrix Scientific

048947-500MG 048947-2.5G
101885-696EA 364.62 USD
101885-696 101885-698
N-(3-Amino-4-fluorophenyl)-3-methylbutanamide
N-(3-Amino-4-fluorophenyl)-3-methylbutanamide

Hazard: Irritant

Formula: C₁₁H₁₅FN₂O
MDL Number: MFCD09049633
CAS Number: 926185-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR