N-(5-Amino-2-fluorophenyl)-3-methylbutanamide

Supplier: Matrix Scientific

048946-500MG 048946-2.5G
101885-692EA 364.62 USD
101885-692 101885-694
N-(5-Amino-2-fluorophenyl)-3-methylbutanamide
N-(5-Amino-2-fluorophenyl)-3-methylbutanamide

Hazard: Irritant

Formula: C₁₁H₁₅FN₂O
MDL Number: MFCD09045134
CAS Number: 926252-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR