N-(5-Amino-2-methylphenyl)-3-methylbutanamide

Supplier: Matrix Scientific

048944-500MG 048944-2.5G
101885-684EA 364.62 USD
101885-684 101885-686
N-(5-Amino-2-methylphenyl)-3-methylbutanamide
N-(5-Amino-2-methylphenyl)-3-methylbutanamide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD09050519
CAS Number: 926193-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR