N-(3-Amino-2-methylphenyl)-3-methylbutanamide

Supplier: Matrix Scientific

048943-500MG 048943-2.5G
101885-680EA 364.62 USD
101885-680 101885-682
N-(3-Amino-2-methylphenyl)-3-methylbutanamide
N-(3-Amino-2-methylphenyl)-3-methylbutanamide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD09043574
CAS Number: 926237-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR