N-(4-Amino-2-methoxyphenyl)-2-methylpropanamide

Supplier: Matrix Scientific

048940-500MG 048940-2.5G
101885-668EA 364.62 USD
101885-668 101885-670
N-(4-Amino-2-methoxyphenyl)-2-methylpropanamide
N-(4-Amino-2-methoxyphenyl)-2-methylpropanamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD03453721
CAS Number: 532417-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR