N-(4-Amino-2-methylphenyl)-2-methylpropanamide
Supplier: Matrix Scientific
048939-500MG
048939-2.5G
101885-664EA
364.62
USD
101885-664
101885-666
N-(4-Amino-2-methylphenyl)-2-methylpropanamide
N-(4-Amino-2-methylphenyl)-2-methylpropanamide
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O |
MDL Number:
MFCD03906874 CAS Number: 766533-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...