N-(5-Amino-2-fluorophenyl)-2-methylpropanamide
Supplier: Matrix Scientific
048934-500MG
048934-2.5G
101885-644EA
364.62
USD
101885-644
101885-646
N-(5-Amino-2-fluorophenyl)-2-methylpropanamide
N-(5-Amino-2-fluorophenyl)-2-methylpropanamide
Hazard: Irritant
Formula:
C₁₀H₁₃FN₂O |
MDL Number:
MFCD09046132 CAS Number: 926259-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...