N-(3-Amino-2-methylphenyl)-2-methylpropanamide

Supplier: Matrix Scientific

048931-500MG 048931-2.5G
101885-632EA 364.62 USD
101885-632 101885-634
N-(3-Amino-2-methylphenyl)-2-methylpropanamide
N-(3-Amino-2-methylphenyl)-2-methylpropanamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD09044678
CAS Number: 926234-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR