N-(4-Amino-2-methoxyphenyl)propanamide

Supplier: Matrix Scientific

048929-500MG 048929-2.5G
101885-624EA 364.62 USD
101885-624 101885-626
N-(4-Amino-2-methoxyphenyl)propanamide
N-(4-Amino-2-methoxyphenyl)propanamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD03906867
CAS Number: 811460-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR