N-(5-Amino-2-chlorophenyl)propanamide

Supplier: Matrix Scientific

048928-500MG 048928-2.5G
101885-620EA 364.62 USD
101885-620 101885-622
N-(5-Amino-2-chlorophenyl)propanamide
N-(5-Amino-2-chlorophenyl)propanamide

Hazard: Irritant

Formula: C₉H₁₁ClN₂O
MDL Number: MFCD11568348
CAS Number: 1082183-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR