N-(4-Amino-2-methoxyphenyl)-3-methoxybenzamide

Supplier: Matrix Scientific

048924-500MG 048924-2.5G
101885-604EA 364.62 USD
101885-604 101885-606
N-(4-Amino-2-methoxyphenyl)-3-methoxybenzamide
N-(4-Amino-2-methoxyphenyl)-3-methoxybenzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₃
MW: 272.31 g/mol
MDL Number: MFCD09046787
CAS Number: 926246-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR