N-(3-Amino-4-chlorophenyl)-3-methoxybenzamide

Supplier: Matrix Scientific

048921-500MG 048921-2.5G
101885-592EA 364.62 USD
101885-592 101885-594
N-(3-Amino-4-chlorophenyl)-3-methoxybenzamide
N-(3-Amino-4-chlorophenyl)-3-methoxybenzamide

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂O₂
MW: 276.72 g/mol
MDL Number: MFCD09045205
CAS Number: 926238-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR