N-(4-Amino-2-chlorophenyl)-4-methoxybenzamide

Supplier: Matrix Scientific

048912-500MG 048912-2.5G
101885-556EA 364.62 USD
101885-556 101885-558
N-(4-Amino-2-chlorophenyl)-4-methoxybenzamide
N-(4-Amino-2-chlorophenyl)-4-methoxybenzamide

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂O₂
MW: 276.72 g/mol
MDL Number: MFCD03906899
CAS Number: 861433-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR