N-(4-Amino-2-methoxyphenyl)-4-methoxybenzamide
Supplier: Matrix Scientific
048911-500MG
048911-2.5G
101885-552EA
364.62
USD
101885-552
101885-554
N-(4-Amino-2-methoxyphenyl)-4-methoxybenzamide
N-(4-Amino-2-methoxyphenyl)-4-methoxybenzamide
Hazard: Irritant
Formula:
C₁₅H₁₆N₂O₃ |
MDL Number:
MFCD03906898 CAS Number: 110506-37-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...