N-(3-Amino-4-methylphenyl)-4-methoxybenzamide

Supplier: Matrix Scientific

048908-500MG 048908-2.5G
101885-540EA 364.62 USD
101885-540 101885-542
N-(3-Amino-4-methylphenyl)-4-methoxybenzamide
N-(3-Amino-4-methylphenyl)-4-methoxybenzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD09936469
CAS Number: 1016796-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR