N-(4-Amino-2-chlorophenyl)-2-phenylacetamide

Supplier: Matrix Scientific

048894-500MG 048894-2.5G
101885-484EA 364.62 USD
101885-484 101885-486
N-(4-Amino-2-chlorophenyl)-2-phenylacetamide
N-(4-Amino-2-chlorophenyl)-2-phenylacetamide

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂O
MDL Number: MFCD03906886
CAS Number: 861224-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR