N-(4-Amino-2-methoxyphenyl)-2-phenylacetamide

Supplier: Matrix Scientific

048893-500MG 048893-2.5G
101885-480EA 364.62 USD
101885-480 101885-482
N-(4-Amino-2-methoxyphenyl)-2-phenylacetamide
N-(4-Amino-2-methoxyphenyl)-2-phenylacetamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD03906885
CAS Number: 869953-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR