N-(5-Amino-2-chlorophenyl)-2-phenylacetamide

Supplier: Matrix Scientific

048892-500MG 048892-2.5G
101885-476EA 364.62 USD
101885-476 101885-478
N-(5-Amino-2-chlorophenyl)-2-phenylacetamide
N-(5-Amino-2-chlorophenyl)-2-phenylacetamide

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂O
MDL Number: MFCD02639536
CAS Number: 428482-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR