N-(3-Amino-4-fluorophenyl)-2-phenylacetamide

Supplier: Matrix Scientific

048891-500MG 048891-2.5G
101885-472EA 364.62 USD
101885-472 101885-474
N-(3-Amino-4-fluorophenyl)-2-phenylacetamide
N-(3-Amino-4-fluorophenyl)-2-phenylacetamide

Hazard: Irritant

Formula: C₁₄H₁₃FN₂O
MDL Number: MFCD09813822
CAS Number: 865837-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR