N-(4-Amino-2-chlorophenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048876-500MG 048876-2.5G
101885-412EA 364.62 USD
101885-412 101885-414
N-(4-Amino-2-chlorophenyl)-2-thiophenecarboxamide
N-(4-Amino-2-chlorophenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₁H₉ClN₂OS
MDL Number: MFCD02189418
CAS Number: 305357-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR