N-(4-Amino-2-methylphenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048875-500MG 048875-2.5G
101885-408EA 364.62 USD
101885-408 101885-410
N-(4-Amino-2-methylphenyl)-2-thiophenecarboxamide
N-(4-Amino-2-methylphenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂OS
MDL Number: MFCD01930151
CAS Number: 351424-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR