N-(3-Amino-4-chlorophenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048874-500MG 048874-2.5G
101885-404EA 364.62 USD
101885-404 101885-406
N-(3-Amino-4-chlorophenyl)-2-thiophenecarboxamide
N-(3-Amino-4-chlorophenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₁H₉ClN₂OS
MDL Number: MFCD09044764
CAS Number: 926203-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR