N-(5-Amino-2-chlorophenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048873-500MG 048873-2.5G
101885-400EA 364.62 USD
101885-400 101885-402
N-(5-Amino-2-chlorophenyl)-2-thiophenecarboxamide
N-(5-Amino-2-chlorophenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₁H₉ClN₂OS
MDL Number: MFCD02258057
CAS Number: 625078-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR