N-(5-Amino-2-methoxyphenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048872-500MG 048872-2.5G
101885-396EA 364.62 USD
101885-396 101885-398
N-(5-Amino-2-methoxyphenyl)-2-thiophenecarboxamide
N-(5-Amino-2-methoxyphenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S
MDL Number: MFCD09044531
CAS Number: 926218-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR