N-(3-Amino-4-fluorophenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048871-500MG 048871-2.5G
101885-392EA 364.62 USD
101885-392 101885-394
N-(3-Amino-4-fluorophenyl)-2-thiophenecarboxamide
N-(3-Amino-4-fluorophenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₁H₉FN₂OS
MDL Number: MFCD09049102
CAS Number: 926251-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR