N-(5-Amino-2-fluorophenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048870-500MG 048870-2.5G
101885-388EA 364.62 USD
101885-388 101885-390
N-(5-Amino-2-fluorophenyl)-2-thiophenecarboxamide
N-(5-Amino-2-fluorophenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₁H₉FN₂OS
MDL Number: MFCD09043023
CAS Number: 926250-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR