N-(3-Amino-4-methylphenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048869-500MG 048869-2.5G
101885-384EA 364.62 USD
101885-384 101885-386
N-(3-Amino-4-methylphenyl)-2-thiophenecarboxamide
N-(3-Amino-4-methylphenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂OS
MDL Number: MFCD02168103
CAS Number: 332906-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR